Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195822.1 | NP_001182751.1 | 3 | 459 | ||
NM_032303.4 | NP_115679.2 | 3 | 459 | |||
NR_036651.1 | 2 | 295 | ||||
XM_011519091.2 | XP_011517393.1 | 3 | 459 | |||
XM_017015203.1 | XP_016870692.1 | 2 | 912 | |||
XM_017015204.1 | XP_016870693.1 | 3 | 1076 | |||
XM_017015205.1 | XP_016870694.1 | 2 | 912 | |||
GenBank mRNA | AK098721.1 | - | 3 | 430 | ||
AK225946.1 | - | 3 | 260 | |||
AK289929.1 | - | 3 | 287 | |||
AK292486.1 | - | 3 | 266 | |||
AK301469.1 | - | 3 | 256 | |||
AK303645.1 | - | 2 | 101 | |||
AK313605.1 | - | 3 | 266 | |||
AY093428.1 | - | 3 | 298 | |||
BC004331.1 | - | 3 | 288 | 3630835 | ||
BC095451.1 | - | 3 | 236 | 30405073 | ||
DB052480.1 | - | 3 | 463 |