Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032142.3 | NP_115518.3 | 19 | 3386 | ||
XM_005258107.3 | XP_005258164.1 | 19 | 3441 | |||
XM_005258109.3 | XP_005258166.1 | 18 | 3231 | |||
XM_005258110.1 | XP_005258167.1 | 6 | 1625 | |||
XM_006722326.3 | XP_006722389.1 | 19 | 3441 | |||
XM_006722327.3 | XP_006722390.1 | 18 | 3231 | |||
XM_006722330.3 | XP_006722393.1 | 13 | 2424 | |||
XM_011525673.2 | XP_011523975.1 | 19 | 3441 | |||
XM_011525675.2 | XP_011523977.1 | 16 | 2990 | |||
XM_017025803.1 | XP_016881292.1 | 18 | 3276 | |||
XM_017025804.1 | XP_016881293.1 | 13 | 2424 | |||
XM_017025805.1 | XP_016881294.1 | 8 | 1765 | |||
XM_017025806.1 | XP_016881295.1 | 16 | 2871 | |||
XM_017025807.1 | XP_016881296.1 | 12 | 2483 | |||
GenBank mRNA | AK074074.1 | - | 4 | 890 | ||
AK096548.1 | - | 16 | 2609 | |||
AL136818.1 | - | 4 | 1039 | |||
AL833494.2 | - | 10 | 2104 | |||
BC144480.1 | - | 9 | 1922 | 9053007 | ||
BC144481.1 | - | 10 | 2132 | 9053008 | ||
BX648232.1 | - | 14 | 2697 | |||
BX649151.1 | - | 6 | 1625 | |||
KJ567064.1 | - | 18 | 3306 |