Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037535.2 | NP_001032624.1 | 4 | 593 | ||
NM_001037536.2 | NP_001032625.1 | 4 | 590 | |||
NM_001037540.2 | NP_001032629.1 | 6 | 823 | |||
NM_006746.5 | NP_006737.2 | 5 | 742 | |||
XM_005274578.2 | XP_005274635.1 | 6 | 818 | |||
XM_005274579.2 | XP_005274636.1 | 5 | 731 | |||
XM_006724508.3 | XP_006724571.1 | 5 | 3723 | |||
XM_006724509.3 | XP_006724572.1 | 4 | 3639 | |||
XM_011545564.2 | XP_011543866.1 | 5 | 3726 | |||
XM_017029723.1 | XP_016885212.1 | 4 | 3642 | |||
GenBank mRNA | AF160728.1 | - | 4 | 530 | ||
AK313993.1 | - | 4 | 551 | |||
BC026159.2 | - | 5 | 722 | 4827364 | ||
BC105028.1 | - | 2 | 199 | 8144031 | ||
BX647757.1 | - | 3 | 2507 |