Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM96A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014812.2 | NP_001014812.1 | 4 | 831 | ||
NM_001289108.1 | NP_001276037.1 | 3 | 3294 | |||
NM_032231.5 | NP_115607.1 | 5 | 881 | |||
NR_110310.1 | 6 | 999 | ||||
GenBank mRNA | BC008865.2 | - | 5 | 783 | 4128736 |