Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256416.2 | NP_001243345.1 | 13 | 1942 | ||
NM_001256417.2 | NP_001243346.1 | 12 | 1768 | |||
NM_032264.4 | NP_115640.1 | 13 | 1978 | |||
NR_046176.2 | 16 | 2507 | ||||
XM_006710957.1 | XP_006711020.1 | 13 | 1836 | |||
XM_006710958.1 | XP_006711021.1 | 16 | 2292 | |||
XM_006710959.2 | XP_006711022.1 | 14 | 2076 | |||
XM_011542276.1 | XP_011540578.1 | 17 | 2549 | |||
XM_011542277.1 | XP_011540579.1 | 17 | 2534 | |||
XM_011542278.1 | XP_011540580.1 | 17 | 2363 | |||
XM_011542281.1 | XP_011540583.1 | 14 | 1899 | |||
XM_017002498.1 | XP_016857987.1 | 17 | 2348 | |||
XM_017002499.1 | XP_016857988.1 | 12 | 1614 | |||
XM_017002500.1 | XP_016857989.1 | 16 | 2367 | |||
XM_017002501.1 | XP_016857990.1 | 17 | 2252 | |||
XM_017002502.1 | XP_016857991.1 | 16 | 2181 | |||
GenBank mRNA | AK095602.1 | - | 16 | 2461 | ||
AK098438.1 | - | 3 | 223 | |||
AK292580.1 | - | 13 | 1843 | |||
AK299840.1 | - | 12 | 1730 | |||
AK314597.1 | - | 13 | 1906 | |||
AL136890.1 | - | 13 | 1755 | |||
AL834330.1 | - | 9 | 951 | |||
AY598326.1 | - | 12 | 1580 | |||
AY894564.1 | - | 12 | 1484 | |||
BC024011.2 | - | 13 | 1869 | 4827023 |