Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136046.2 | NP_001129518.1 | 14 | 2383 | ||
NM_001267822.1 | NP_001254751.1 | 14 | 2407 | |||
NM_032265.2 | NP_115641.1 | 13 | 2266 | |||
XM_017025218.1 | XP_016880707.1 | 11 | 1822 | |||
GenBank mRNA | AK302181.1 | - | 14 | 2374 | ||
AL136893.1 | - | 12 | 2239 |