Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304353.1 | NP_001291282.1 | 3 | 254 | ||
NM_001304354.1 | NP_001291283.1 | 3 | 251 | |||
NM_032309.3 | NP_115685.1 | 3 | 417 | |||
XM_005263824.3 | XP_005263881.1 | 3 | 1248 | |||
GenBank mRNA | AK096990.1 | - | 3 | 259 | ||
AK312219.1 | - | 3 | 393 | |||
BC004498.2 | - | 3 | 395 | 3831856 | ||
BM563093.1 | - | 3 | 251 | |||
BX115079.1 | - | 3 | 253 | |||
HY011716.1 | - | 3 | 412 | |||
N39730.1 | - | 3 | 252 |