Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXRA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282582.1 | NP_001269511.1 | 9 | 1225 | ||
NM_001282583.1 | NP_001269512.1 | 8 | 1644 | |||
NM_001282584.1 | NP_001269513.1 | 7 | 1144 | |||
NM_001282585.1 | NP_001269514.1 | 8 | 1200 | |||
NM_032348.3 | NP_115724.1 | 8 | 1447 | |||
XM_017001516.1 | XP_016857005.1 | 8 | 1224 | |||
XM_017001517.1 | XP_016857006.1 | 7 | 2851 | |||
GenBank mRNA | AB052096.1 | - | 9 | 1225 | ||
AK027269.1 | - | 8 | 1200 | |||
AK095966.1 | - | 8 | 1644 | |||
AK225028.1 | - | 8 | 1186 | |||
AK293442.1 | - | 7 | 913 | |||
AK304615.1 | - | 7 | 1239 | |||
AK315510.1 | - | 8 | 1214 | |||
AL050202.1 | - | 2 | 83 | |||
AY358391.1 | - | 7 | 890 | |||
BC006213.2 | - | 8 | 1188 | 3343261 | ||
BC017312.2 | - | 8 | 1188 | 5090754 |