Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032354.3 | NP_115730.2 | 5 | 629 | ||
NM_183065.2 | NP_898888.1 | 5 | 611 | |||
GenBank mRNA | AF311338.1 | - | 5 | 685 | ||
AK027813.1 | - | 3 | 1975 | |||
AK127891.1 | - | 2 | 2066 | |||
AY358525.1 | - | 5 | 589 | |||
BC006292.2 | - | 2 | 302 | |||
BC070231.1 | - | 5 | 600 | 6617879 | ||
BM920977.1 | - | 5 | 582 |