Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKT1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098632.2 | NP_001092102.1 | 2, 3 | 757 | ||
NM_001098633.3 | NP_001092103.1 | 2, 3 | 545 | |||
NM_001278159.1 | NP_001265088.1 | 2, 3 | 770 | |||
NM_001278160.1 | NP_001265089.1 | 2, 3 | 581 | |||
NM_032375.5 | NP_115751.3 | 2, 3 | 1179 | |||
GenBank mRNA | AK055511.1 | - | 2, 3 | 770 | ||
AK092610.1 | - | 1, 2 | 1114 | |||
AK123687.1 | - | 2, 3 | 1144 | |||
AK226004.1 | - | 2, 3 | 1070 | |||
AK316603.1 | - | 2, 3 | 411 | |||
BC007416.2 | - | 2, 3 | 1065 | 2988648 | ||
BC015562.1 | - | 3, 4 | 349 | |||
BC016043.1 | - | 2, 3 | 565 | 4635389 | ||
BC051844.2 | - | 2, 3 | 649 | 6164197 | ||
DA542314.1 | - | 2, 3 | 513 |