Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304813.1 | NP_001291742.1 | 4 | 591 | ||
NM_001304814.1 | NP_001291743.1 | 4 | 688 | |||
NM_032376.3 | NP_115752.1 | 3 | 520 | |||
XM_005257737.2 | XP_005257794.1 | 4 | 1868 | |||
XM_011525346.1 | XP_011523648.1 | 6 | 2105 | |||
XM_011525347.1 | XP_011523649.1 | 5 | 1982 | |||
XM_011525349.1 | XP_011523651.1 | 4 | 1888 | |||
XM_011525351.1 | XP_011523653.1 | 4 | 1335 | |||
XM_011525353.1 | XP_011523655.1 | 4 | 1747 | |||
GenBank mRNA | AB097008.1 | - | 3 | 456 | ||
AK130810.1 | - | 3 | 454 | |||
AK172826.1 | - | 3 | 472 | |||
AK294237.1 | - | 3 | 371 | |||
AK313855.1 | - | 3 | 454 | |||
BC007438.2 | - | 3 | 468 | 3028940 | ||
CN261562.1 | - | 3 | 521 | |||
DC311483.1 | - | 3 | 519 |