Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007136.3 | NP_009067.2 | 1 | 578 | ||
XM_017007132.1 | XP_016862621.1 | 1 | 266 | |||
XM_017007133.1 | XP_016862622.1 | 1 | 266 | |||
XM_017007134.1 | XP_016862623.1 | 1 | 266 | |||
XM_017007135.1 | XP_016862624.1 | 1 | 266 | |||
XM_017007136.1 | XP_016862625.1 | 1 | 266 | |||
GenBank mRNA | BC069606.1 | - | 1 | 272 | 7262407 | |
BC069826.1 | - | 1 | 271 | 7262419 | ||
BC129925.1 | - | 1 | 101 | 40121991 | ||
X65233.1 | - | 1 | 586 |