Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM120B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286379.1 | NP_001273308.1 | 2 | 684 | ||
NM_001286380.1 | NP_001273309.1 | 2 | 447 | |||
NM_032448.2 | NP_115824.1 | 2 | 435 | |||
XM_011536185.2 | XP_011534487.1 | 2 | 429 | |||
XM_017011358.1 | XP_016866847.1 | 2 | 429 | |||
XM_017011359.1 | XP_016866848.1 | 2 | 429 | |||
XM_017011360.1 | XP_016866849.1 | 2 | 429 | |||
GenBank mRNA | AB058741.1 | - | 2 | 348 | ||
AK056435.1 | - | 2 | 415 | |||
AK294415.1 | - | 2 | 684 | |||
AK299890.1 | - | 2 | 447 | |||
AL832883.1 | - | 2 | 333 | |||
BC012177.1 | - | 2 | 401 | 4650497 | ||
DQ873695.1 | - | 2 | 436 |