Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199743.1 | NP_001186672.1 | 3 | 394 | ||
NM_032486.3 | NP_115875.1 | 3 | 394 | |||
NR_037573.1 | 4 | 495 | ||||
GenBank mRNA | AK027387.1 | - | 3 | 257 | ||
AK292843.1 | - | 3 | 321 | |||
AK299631.1 | - | 3 | 291 | |||
BC004191.2 | - | 3 | 274 | 3505855 | ||
CD388246.1 | - | 3 | 313 | |||
DA310703.1 | - | 3 | 394 | |||
DB087566.1 | - | 4 | 351 |