Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033002.3 | NP_001028174.2 | 2 | 767 | ||
NM_001160243.1 | NP_001153715.1 | 2 | 767 | |||
NM_001160244.1 | NP_001153716.1 | 2 | 767 | |||
NM_001160246.1 | NP_001153718.1 | 2 | 767 | |||
NM_001160266.1 | NP_001153738.1 | 2 | 767 | |||
NR_027679.1 | 2 | 767 | ||||
NR_027682.1 | 2 | 767 | ||||
NR_027683.1 | 2 | 767 | ||||
NR_027684.1 | 2 | 767 | ||||
NR_027685.1 | 2 | 767 | ||||
GenBank mRNA | AK096001.1 | - | 2 | 223 | ||
AK226044.1 | - | 2 | 523 | |||
AK295394.1 | - | 2 | 208 | |||
AK300409.1 | - | 2 | 223 | |||
AY680654.1 | - | 2 | 197 | |||
AY680655.1 | - | 2 | 197 | |||
AY680656.1 | - | 2 | 197 | |||
AY680657.1 | - | 2 | 197 | |||
AY680658.1 | - | 2 | 197 | |||
AY680659.1 | - | 2 | 197 | |||
AY680660.1 | - | 2 | 197 | |||
AY775314.1 | - | 2 | 767 | |||
AY775315.1 | - | 2 | 767 | |||
AY775316.1 | - | 2 | 767 | |||
AY775317.1 | - | 2 | 767 | |||
AY775318.1 | - | 2 | 767 | |||
AY775319.1 | - | 2 | 767 | |||
AY775320.1 | - | 2 | 767 | |||
AY775321.1 | - | 2 | 767 | |||
AY775323.1 | - | 2 | 767 | |||
BC004451.1 | - | 2 | 210 | 2820882 | ||
BC013831.1 | - | 2 | 238 | |||
BC046349.1 | - | 2 | 215 | |||
BC051849.2 | - | 2 | 200 | 5493799 | ||
BI091755.1 | - | 2 | 203 | |||
CK005117.1 | - | 2 | 226 | |||
DA349558.1 | - | 2 | 206 | |||
N41653.1 | - | 2 | 165 |