Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172648.3 | NP_001166119.1 | 11 | 1728 | ||
NM_001172649.3 | NP_001166120.1 | 10 | 1825 | |||
NM_001291106.1 | NP_001278035.1 | 11 | 1972 | |||
NM_001291107.1 | NP_001278036.1 | 11 | 1972 | |||
NM_007185.6 | NP_009116.3 | 11 | 1975 | |||
XM_006711139.2 | XP_006711202.1 | 12 | 1203 | |||
XM_006711141.2 | XP_006711204.1 | 12 | 1200 | |||
XM_011509111.2 | XP_011507413.1 | 10 | 1670 | |||
XM_011509112.2 | XP_011507414.1 | 10 | 1066 | |||
XM_011509114.2 | XP_011507416.1 | 10 | 1034 | |||
XM_017000145.1 | XP_016855634.1 | 10 | 1041 | |||
XM_017000146.1 | XP_016855635.1 | 10 | 1038 | |||
GenBank mRNA | AB209264.1 | - | 11 | 1728 | ||
AF284423.1 | - | 2 | 202 | |||
AF329264.1 | - | 11 | 1166 | |||
AK298852.1 | - | 5 | 598 | |||
AY165003.1 | - | 11 | 1965 | |||
BC041088.1 | - | 10 | 1057 | |||
BC052491.1 | - | 11 | 1946 | 4820737 | ||
BC068008.1 | - | 10 | 1002 | |||
BC104758.1 | - | 11 | 1238 | 8143761 | ||
BC143226.1 | - | 11 | 1235 | 9051727 | ||
U80746.1 | - | 8 | 854 | |||
U80759.1 | - | 8 | 861 |