Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184992.1 | NP_001171921.1 | 6 | 1096 | ||
NM_032322.3 | NP_115698.3 | 5 | 1031 | |||
NM_197939.1 | NP_922921.1 | 4 | 868 | |||
XM_005258043.4 | XP_005258100.1 | 6 | 2543 | |||
XM_017025223.1 | XP_016880712.1 | 5 | 1217 | |||
GenBank mRNA | AB470605.1 | - | 5 | 895 | ||
AK122646.1 | - | 4 | 741 | |||
AK122709.1 | - | 6 | 826 | |||
AK226030.1 | - | 3 | 614 | |||
BC005084.1 | - | 4 | 783 | |||
BC082262.1 | - | 6 | 974 | |||
BC126420.1 | - | 5 | 985 | 8992136 | ||
BC126422.1 | - | 5 | 985 | 8992138 | ||
BC144143.1 | - | 3 | 678 |