Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032329.4 | NP_115705.2 | 4 | 369 | ||
XM_005247047.2 | XP_005247104.1 | 4 | 413 | |||
XM_017005097.1 | XP_016860586.1 | 4 | 1437 | |||
XM_017005098.1 | XP_016860587.1 | 4 | 1437 | |||
XM_017005099.1 | XP_016860588.1 | 4 | 1437 | |||
XM_017005100.1 | XP_016860589.1 | 5 | 2169 | |||
XM_017005101.1 | XP_016860590.1 | 5 | 2169 | |||
XM_017005102.1 | XP_016860591.1 | 4 | 413 | |||
GenBank mRNA | AF189286.1 | - | 4 | 346 | ||
AK074422.1 | - | 4 | 369 | |||
AK289958.1 | - | 4 | 370 | |||
AK300790.1 | - | 4 | 368 | |||
BC005370.1 | - | 4 | 365 | 3932127 | ||
BC071899.1 | - | 4 | 360 | 6111003 |