Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032339.4 | NP_115715.3 | 4 | 574 | ||
XM_005257736.3 | XP_005257793.1 | 3 | 632 | |||
GenBank mRNA | AY508814.1 | - | 4 | 574 | ||
BC006006.1 | - | 4 | 513 | 4283597 | ||
BC106016.1 | - | 2 | 721 | |||
BX646129.1 | - | 4 | 547 |