Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SP140 SIRNA Assays ›
Gene Symbol:
SP140
Gene Name:
SP140 nuclear body protein
Gene Aliases:
LYSP100, LYSP100-A, LYSP100-B
Chromosome Location:
Chr.2: 230202924 - 230316571 on Build GRCh38
UniGene ID:
Hs.632549
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
3413
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SP140
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005176.2
NP_001005176.1
2
254
NM_001278451.1
NP_001265380.1
2
254
NM_001278452.1
NP_001265381.1
2
254
NM_001278453.1
NP_001265382.1
2
254
NM_007237.4
NP_009168.4
2
254
XM_005246252.2
XP_005246309.1
2
218
XM_005246253.2
XP_005246310.1
2
218
XM_005246254.2
XP_005246311.1
2
218
XM_005246255.2
XP_005246312.1
2
218
XM_005246256.2
XP_005246313.1
2
218
XM_006712223.3
XP_006712286.1
2
218
XM_011510515.1
XP_011508817.1
2
218
XM_011510516.1
XP_011508818.1
2
218
XM_011510517.2
XP_011508819.1
4
1082
XM_011510518.2
XP_011508820.1
3
354
XM_011510519.1
XP_011508821.1
2
218
XM_011510520.1
XP_011508822.1
2
218
XM_017003239.1
XP_016858728.1
2
218
XM_017003240.1
XP_016858729.1
2
218
XM_017003241.1
XP_016858730.1
2
218
XM_017003242.1
XP_016858731.1
2
218
XM_017003243.1
XP_016858732.1
2
218
XM_017003244.1
XP_016858733.1
3
354
XM_017003245.1
XP_016858734.1
2
218
XM_017003246.1
XP_016858735.1
2
218
XM_017003247.1
XP_016858736.1
2
218
XM_017003248.1
XP_016858737.1
2
218
XM_017003249.1
XP_016858738.1
2
218
XM_017003250.1
XP_016858739.1
2
218
XM_017003251.1
XP_016858740.1
3
361
XM_017003252.1
XP_016858741.1
2
218
GenBank mRNA
AK301267.1
-
2
254
AK301317.1
-
2
255
BC038091.1
-
2
225
BC054890.1
-
2
224
BC070160.1
-
2
220
BC105743.1
-
2
237
40035747
BC105744.1
-
2
239
40035746
BC105960.1
-
2
239
40035752
DC391925.1
-
2
254
U36499.1
-
2
255
U36500.1
-
2
255
U63420.1
-
2
245
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
defense response
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0