Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXRA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282582.1 | NP_001269511.1 | 9, 10 | 1285 | ||
NM_001282583.1 | NP_001269512.1 | 8, 9 | 1704 | |||
NM_001282584.1 | NP_001269513.1 | 7, 8 | 1204 | |||
NM_032348.3 | NP_115724.1 | 8, 9 | 1507 | |||
XM_017001516.1 | XP_016857005.1 | 8, 9 | 1284 | |||
XM_017001517.1 | XP_016857006.1 | 7, 8 | 2911 | |||
GenBank mRNA | AB052096.1 | - | 9, 10 | 1285 | ||
AK095966.1 | - | 8, 9 | 1704 | |||
AK225028.1 | - | 8, 9 | 1246 | |||
AK293442.1 | - | 7, 8 | 973 | |||
AK304615.1 | - | 7, 8 | 1299 | |||
AK315510.1 | - | 8, 9 | 1274 | |||
AL050202.1 | - | 2, 3 | 143 | |||
AY358391.1 | - | 7, 8 | 950 | |||
BC006213.2 | - | 8, 9 | 1248 | 3343261 | ||
BC017312.2 | - | 8, 9 | 1248 | 5090754 |