Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKT1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098632.2 | NP_001092102.1 | 5 | 1405 | ||
NM_001098633.3 | NP_001092103.1 | 5 | 1193 | |||
NM_001278159.1 | NP_001265088.1 | 5 | 1418 | |||
NM_001278160.1 | NP_001265089.1 | 5 | 1229 | |||
NM_032375.5 | NP_115751.3 | 5 | 1827 | |||
GenBank mRNA | AK055511.1 | - | 5 | 1418 | ||
AK092610.1 | - | 4 | 1762 | |||
AK123687.1 | - | 5 | 1792 | |||
BC000031.2 | - | 3 | 622 | |||
BC007416.2 | - | 5 | 1713 | 2988648 | ||
BC015562.1 | - | 6 | 997 | |||
BC051844.2 | - | 5 | 1297 | 6164197 |