Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032383.4 | NP_115759.2 | 2 | 441 | ||
XM_005247834.3 | XP_005247891.1 | 2 | 416 | |||
XM_017007323.1 | XP_016862812.1 | 2 | 416 | |||
GenBank mRNA | AK055245.1 | - | 2 | 900 | ||
AK291631.1 | - | 2 | 416 | |||
AY033141.1 | - | 2 | 441 | |||
BC040359.1 | - | 2 | 378 | 4385940 |