Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032389.4 | NP_115765.2 | 5 | 730 | ||
XM_005253166.1 | XP_005253223.1 | 5 | 1108 | |||
XM_005253167.1 | XP_005253224.1 | 5 | 1108 | |||
XM_005253168.2 | XP_005253225.1 | 5 | 1108 | |||
XM_006718346.1 | XP_006718409.1 | 5 | 1108 | |||
XM_017018413.1 | XP_016873902.1 | 5 | 1108 | |||
XM_017018414.1 | XP_016873903.1 | 5 | 504 | |||
XM_017018415.1 | XP_016873904.1 | 5 | 498 | |||
GenBank mRNA | AB073358.1 | - | 4 | 340 | ||
AK027482.1 | - | 5 | 466 | |||
AK300347.1 | - | 5 | 464 | |||
AL834337.1 | - | 5 | 449 | |||
BC030148.2 | - | 5 | 473 | 4558453 | ||
BC036823.1 | - | 5 | 475 |