Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARDBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007375.3 | NP_031401.1 | 2 | 273 | ||
XM_017000863.1 | XP_016856352.1 | 2 | 261 | |||
XM_017000864.1 | XP_016856353.1 | 2 | 261 | |||
XM_017000865.1 | XP_016856354.1 | 2 | 261 | |||
XM_017000866.1 | XP_016856355.1 | 2 | 261 | |||
XM_017000867.1 | XP_016856356.1 | 2 | 261 | |||
XM_017000868.1 | XP_016856357.1 | 2 | 261 | |||
GenBank mRNA | AK222754.1 | - | 2 | 211 | ||
AK295920.1 | - | 2 | 211 | |||
AK312416.1 | - | 2 | 241 | |||
AL050265.1 | - | 2 | 227 | |||
BC001487.2 | - | 2 | 226 | |||
BC071657.1 | - | 2 | 224 | 5498250 | ||
BC095435.1 | - | 2 | 225 | 30389805 | ||
CD703666.1 | - | 2 | 299 | |||
CR533534.1 | - | 1 | 139 | |||
EF434181.1 | - | 1 | 139 | |||
EF434182.1 | - | 1 | 139 | |||
EF434183.1 | - | 1 | 139 | |||
HQ628633.1 | - | 1 | 139 | |||
HQ628635.1 | - | 1 | 139 | |||
HQ628636.1 | - | 1 | 139 | |||
HQ628637.1 | - | 1 | 139 | |||
U23731.1 | - | 1 | 139 |