Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271286.1 | NP_001258215.1 | 3 | 263 | ||
NM_001271287.1 | NP_001258216.1 | 2 | 207 | |||
NM_001271288.1 | NP_001258217.1 | 2 | 204 | |||
NM_001271289.1 | NP_001258218.1 | 3 | 252 | |||
NM_001271318.1 | NP_001258247.1 | 4 | 367 | |||
NM_032434.3 | NP_115810.2 | 3 | 266 | |||
GenBank mRNA | AB058708.1 | - | 1 | 25 | ||
AK057028.1 | - | 3 | 176 | |||
AK293771.1 | - | 3 | 166 | |||
AK294055.1 | - | 3 | 372 | |||
AK299818.1 | - | 3 | 153 | |||
AK316011.1 | - | 4 | 227 | |||
AK316299.1 | - | 2 | 121 | |||
AL514772.3 | - | 3 | 183 | |||
BC043221.2 | - | 2 | 115 | 5295402 | ||
BC093041.1 | - | 2 | 134 | 6173398 | ||
BG720805.1 | - | 3 | 216 | |||
BP251717.1 | - | 3 | 266 | |||
BX647996.1 | - | 2 | 113 |