Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032476.3 | NP_115865.1 | 3 | 457 | ||
GenBank mRNA | AB049942.1 | - | 3 | 403 | ||
AK096374.1 | - | 3 | 1424 | |||
AK312084.1 | - | 3 | 402 | |||
AY061855.1 | - | 3 | 457 | |||
BC000547.2 | - | 3 | 389 | 3162515 | ||
BC004976.1 | - | 3 | 391 | 2958115 | ||
BC010076.2 | - | 3 | 391 | 4304420 | ||
BC051302.1 | - | 3 | 298 | 4855355 | ||
BG720377.1 | - | 3 | 407 |