Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032478.3 | NP_115867.2 | 9 | 1514 | ||
GenBank mRNA | AF161380.1 | - | 5 | 673 | ||
BC013311.2 | - | 9 | 1313 | 3546083 |