Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281520.1 | NP_001268449.1 | 2, 3 | 572 | ||
NM_001281521.1 | NP_001268450.1 | 2, 3 | 518 | |||
NM_001281522.1 | NP_001268451.1 | 2, 3 | 518 | |||
NM_032485.5 | NP_115874.3 | 2, 3 | 518 | |||
NM_182802.2 | NP_877954.1 | 2, 3 | 572 | |||
XM_011529387.2 | XP_011527689.1 | 2, 3 | 481 | |||
XM_017028105.1 | XP_016883594.1 | 2, 3 | 547 | |||
GenBank mRNA | AJ439063.1 | - | 2, 3 | 518 | ||
AK297806.1 | - | 2, 3 | 451 | |||
AK314654.1 | - | 2, 3 | 200 | |||
AY158211.1 | - | 2, 3 | 271 | |||
BC005170.1 | - | 2, 3 | 271 | |||
BC080656.1 | - | 2, 3 | 223 | 4841271 | ||
BC101054.1 | - | 2, 3 | 319 | 40008094 | ||
BC101055.1 | - | 2, 3 | 319 | 40008095 | ||
BC101056.1 | - | 2, 3 | 319 | 40008096 | ||
BC101057.1 | - | 2, 3 | 373 | 40008097 |