Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012118.3 | NP_036250.2 | 3 | 703 | ||
GenBank mRNA | AF183961.1 | - | 3 | 673 | ||
AF199493.1 | - | 2 | 156 | |||
AF199494.1 | - | 1 | 526 | |||
BC113494.1 | - | 3 | 567 | 8322546 | ||
BC113500.1 | - | 3 | 567 | 8322552 |