Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184992.1 | NP_001171921.1 | 6 | 1509 | ||
NM_032322.3 | NP_115698.3 | 5 | 1444 | |||
NM_197939.1 | NP_922921.1 | 4 | 1281 | |||
XM_005258043.4 | XP_005258100.1 | 6 | 2956 | |||
XM_017025223.1 | XP_016880712.1 | 5 | 1630 | |||
GenBank mRNA | AJ496729.1 | - | 5 | 1359 | ||
AK122646.1 | - | 4 | 1154 | |||
AK122709.1 | - | 6 | 1239 | |||
BC005084.1 | - | 4 | 1196 | |||
BC082262.1 | - | 6 | 1387 | |||
BC126420.1 | - | 5 | 1398 | 8992136 | ||
BC126422.1 | - | 5 | 1398 | 8992138 | ||
BC144143.1 | - | 3 | 1091 |