Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTPCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF416713.1 | - | 4 | 543 | ||
AK316591.1 | - | 4 | 633 | |||
BC005102.1 | - | 4 | 581 | 3619026 | ||
BF663221.1 | - | 2 | 311 | |||
BG106026.1 | - | 3 | 419 | |||
BX537541.1 | - | 1 | 8208 | |||
W80669.1 | - | 3 | 401 |