Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297423.1 | NP_001284352.1 | 11 | 1979 | ||
NM_001297424.1 | NP_001284353.1 | 9 | 1295 | |||
NM_001297425.1 | NP_001284354.1 | 9 | 1292 | |||
NM_001297426.1 | NP_001284355.1 | 12 | 1759 | |||
NM_001297427.1 | NP_001284356.1 | 11 | 2015 | |||
NM_001297428.1 | NP_001284357.1 | 9 | 1331 | |||
NM_032326.3 | NP_115702.1 | 11 | 1518 | |||
XM_005272301.2 | XP_005272358.1 | 10 | 1409 | |||
XM_005272303.4 | XP_005272360.1 | 12 | 1675 | |||
XM_005272304.1 | XP_005272361.1 | 11 | 1484 | |||
XM_011513576.1 | XP_011511878.1 | 7 | 1036 | |||
XM_017008701.1 | XP_016864190.1 | 12 | 1595 | |||
XM_017008702.1 | XP_016864191.1 | 9 | 1242 | |||
XM_017008703.1 | XP_016864192.1 | 12 | 2037 | |||
XM_017008704.1 | XP_016864193.1 | 10 | 1850 | |||
XM_017008705.1 | XP_016864194.1 | 8 | 1187 | |||
GenBank mRNA | AL834199.1 | - | 12 | 1629 | ||
BC005158.2 | - | 11 | 1431 | 3505819 | ||
BC118552.1 | - | 1 | 338 |