Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZDHHC16 SIRNA Assays ›
Gene Symbol:
ZDHHC16
Gene Name:
zinc finger DHHC-type containing 16
Gene Aliases:
APH2, DHHC-16
Chromosome Location:
Chr.10: 97446131 - 97457370 on Build GRCh38
UniGene ID:
Hs.76662
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
34207
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZDHHC16
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001287803.1
NP_001274732.1
10, 11
1304
NM_001287804.1
NP_001274733.1
6, 7
968
NM_032327.3
NP_115703.2
9, 10
1178
NM_198043.2
NP_932160.1
8, 9
1130
NM_198044.2
NP_932161.1
8, 9
1061
NM_198045.2
NP_932162.1
7, 8
935
NM_198046.2
NP_932163.1
10, 11
1358
NR_109895.1
7, 8
681
XM_006718021.1
XP_006718084.1
9, 10
1187
XM_006718022.1
XP_006718085.1
10, 11
1290
XM_006718023.1
XP_006718086.1
8, 9
1139
XM_006718024.1
XP_006718087.1
9, 10
1112
XM_006718025.1
XP_006718088.1
8, 9
1070
XM_006718026.1
XP_006718089.1
7, 8
1022
XM_017016766.1
XP_016872255.1
9, 10
1221
XM_017016767.1
XP_016872256.1
8, 9
1064
XM_017016768.1
XP_016872257.1
9, 10
1152
XM_017016769.1
XP_016872258.1
7, 8
1001
XM_017016770.1
XP_016872259.1
8, 9
957
XM_017016771.1
XP_016872260.1
7, 8
1059
XM_017016772.1
XP_016872261.1
7, 8
909
XM_017016773.1
XP_016872262.1
10, 11
1378
XM_017016774.1
XP_016872263.1
10, 11
1357
GenBank mRNA
AB209272.1
-
7, 8
2884
AF258563.1
-
8, 9
1059
AJ294945.1
-
10, 11
1192
AK074427.1
-
9, 10
1132
AK297962.1
-
10, 11
1260
AY359080.1
-
8, 9
1021
BC004535.2
-
7, 8
856
3160750
BC008074.2
-
9, 10
1136
3029918
BC011749.2
-
9, 10
1124
3140177
BC012830.2
-
9, 10
1136
3029918
BC033157.1
-
8, 9
988
4309312
BG769550.1
-
7, 8
618
BX353685.2
-
6, 7
904
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
protein palmitoylation
zinc ion binding
palmitoyltransferase activity
protein-cysteine S-palmitoyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0