Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099737.2 | NP_001093207.1 | 10 | 1058 | ||
NM_032332.3 | NP_115708.1 | 8 | 801 | |||
NR_029375.1 | 10 | 1041 | ||||
GenBank mRNA | AK074525.1 | - | 10 | 1031 | ||
AK223195.1 | - | 8 | 751 | |||
AK314164.1 | - | 10 | 1035 | |||
BC005870.2 | - | 8 | 734 | 2961616 |