Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM213A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243778.1 | NP_001230707.1 | 6 | 968 | ||
NM_001243779.1 | NP_001230708.1 | 6 | 1038 | |||
NM_001243780.1 | NP_001230709.1 | 6 | 1534 | |||
NM_001243781.1 | NP_001230710.1 | 6 | 1051 | |||
NM_001243782.1 | NP_001230711.1 | 6 | 951 | |||
NM_032333.4 | NP_115709.3 | 6 | 1068 | |||
XM_011540265.2 | XP_011538567.1 | 6 | 1371 | |||
XM_011540266.2 | XP_011538568.1 | 6 | 1774 | |||
XM_017016775.1 | XP_016872264.1 | 6 | 1055 | |||
GenBank mRNA | AF086462.1 | - | 314 | |||
AF305824.1 | - | 4 | 723 | |||
AK074643.1 | - | 6 | 1013 | |||
AK075447.1 | - | 6 | 1043 | |||
AY359059.1 | - | 6 | 961 | |||
BC005871.2 | - | 6 | 930 | 3010078 |