Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130146.1 | NP_001123618.1 | 3, 4 | 399 | ||
NM_001130147.1 | NP_001123619.1 | 3, 4 | 426 | |||
NM_001130148.1 | NP_001123620.1 | 2, 3 | 372 | |||
NM_032358.3 | NP_115734.1 | 4, 5 | 453 | |||
GenBank mRNA | AK027638.1 | - | 4, 5 | 371 | ||
AK293165.1 | - | 3, 4 | 337 | |||
AK308216.1 | - | 3, 4 | 381 | |||
AK316029.1 | - | 2, 3 | 374 | |||
BC006444.2 | - | 3, 4 | 342 | 3616619 | ||
CX784327.1 | - | 3, 4 | 306 | |||
DA711064.1 | - | 4, 5 | 454 | |||
DC331633.1 | - | 2, 3 | 372 |