Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242832.1 | NP_001229761.1 | 9 | 1060 | ||
NM_032389.4 | NP_115765.2 | 10 | 1144 | |||
XM_005253166.1 | XP_005253223.1 | 11 | 1639 | |||
XM_005253167.1 | XP_005253224.1 | 11 | 1606 | |||
XM_005253168.2 | XP_005253225.1 | 11 | 1564 | |||
XM_006718346.1 | XP_006718409.1 | 10 | 1597 | |||
XM_011520405.1 | XP_011518707.1 | 10 | 1480 | |||
XM_017018413.1 | XP_016873902.1 | 10 | 1564 | |||
XM_017018414.1 | XP_016873903.1 | 10 | 960 | |||
XM_017018415.1 | XP_016873904.1 | 10 | 912 | |||
GenBank mRNA | AB073358.1 | - | 9 | 754 | ||
AK027482.1 | - | 10 | 880 | |||
AK098324.1 | - | 5 | 490 | |||
AK301785.1 | - | 9 | 764 | |||
AK304541.1 | - | 10 | 719 | |||
AK315992.1 | - | 4 | 411 | |||
AL834337.1 | - | 10 | 863 | |||
BC030148.2 | - | 10 | 887 | 4558453 | ||
BC036823.1 | - | 10 | 889 |