Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271286.1 | NP_001258215.1 | 5, 6 | 647 | ||
NM_001271287.1 | NP_001258216.1 | 5, 6 | 675 | |||
NM_001271288.1 | NP_001258217.1 | 5, 6 | 672 | |||
NM_001271289.1 | NP_001258218.1 | 5, 6 | 636 | |||
NM_001271318.1 | NP_001258247.1 | 7, 8 | 835 | |||
NM_032434.3 | NP_115810.2 | 6, 7 | 734 | |||
GenBank mRNA | AB058708.1 | - | 4, 5 | 493 | ||
AK057028.1 | - | 6, 7 | 644 | |||
AK293771.1 | - | 5, 6 | 550 | |||
AK294055.1 | - | 6, 7 | 840 | |||
AK299818.1 | - | 5, 6 | 537 | |||
AK316011.1 | - | 7, 8 | 695 | |||
AK316299.1 | - | 5, 6 | 589 | |||
AL514772.3 | - | 6, 7 | 651 | |||
BC043221.2 | - | 5, 6 | 583 | 5295402 | ||
BC093041.1 | - | 5, 6 | 602 | 6173398 | ||
BX647996.1 | - | 5, 6 | 581 |