Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142623.1 | NP_001136095.1 | 10 | 2358 | ||
NM_032484.4 | NP_115873.1 | 10 | 2269 | |||
GenBank mRNA | AK074700.1 | - | 9 | 2473 | ||
AK075277.1 | - | 10 | 2320 | |||
AK122777.1 | - | 11 | 2037 | |||
AK123075.1 | - | 2287 | ||||
BC022784.1 | - | 10 | 2204 | 3847323 |