Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199743.1 | NP_001186672.1 | 6 | 1707 | ||
NM_032486.3 | NP_115875.1 | 6 | 1677 | |||
NR_037573.1 | 7 | 1778 | ||||
GenBank mRNA | AK027387.1 | - | 6 | 1540 | ||
AK126469.1 | - | 1 | 1628 | |||
BC004191.2 | - | 6 | 1557 | 3505855 |