Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032494.2 | NP_115883.2 | 3, 4 | 483 | ||
XM_011511998.1 | XP_011510300.1 | 3, 4 | 428 | |||
XM_017005107.1 | XP_016860596.1 | 3, 4 | 436 | |||
GenBank mRNA | AF334161.1 | - | 3, 4 | 469 | ||
AK315329.1 | - | 3, 4 | 437 | |||
BC032001.1 | - | 3, 4 | 421 | 4749794 | ||
CV574571.1 | - | 1, 2 | 29 |