Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM185A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174092.2 | NP_001167563.1 | 2 | 386 | ||
NM_001282302.1 | NP_001269231.1 | 3 | 563 | |||
NM_032508.3 | NP_115897.1 | 3 | 563 | |||
GenBank mRNA | AF530473.1 | - | 4 | 542 | ||
AK096308.1 | - | 3 | 375 | |||
AK297335.1 | - | 2 | 249 | |||
BC103820.1 | - | 3 | 267 | |||
BC103821.1 | - | 3 | 267 | 40000939 | ||
BC103822.1 | - | 3 | 267 | 40000940 | ||
BG721461.1 | - | 2 | 162 | |||
BQ224383.1 | - | 3 | 354 |