Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032511.2 | NP_115900.1 | 1 | 390 | ||
XM_011536189.2 | XP_011534491.1 | 1 | 370 | |||
GenBank mRNA | AK096480.1 | - | 1 | 153 | ||
BC004869.2 | - | 1 | 390 | 3834272 | ||
BC011583.1 | - | 1 | 390 | 4577456 |