Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303524.1 | NP_001290453.1 | 5 | 1343 | ||
NM_001303526.1 | NP_001290455.1 | 3 | 1334 | |||
NM_001303527.1 | NP_001290456.1 | 3 | 1336 | |||
NM_001303528.1 | NP_001290457.1 | 5 | 1561 | |||
NM_001303529.1 | NP_001290458.1 | 4 | 1703 | |||
NM_001303530.1 | NP_001290459.1 | 5 | 1912 | |||
NM_032525.2 | NP_115914.1 | 4 | 1445 | |||
GenBank mRNA | AK001295.1 | - | 4 | 1263 | ||
AK022340.1 | - | 4 | 1263 | |||
AK092677.1 | - | 3 | 1600 | |||
AK098440.1 | - | 4 | 1263 | |||
AK298196.1 | - | 5 | 1159 | |||
AK304616.1 | - | 5 | 1145 | |||
BC002654.1 | - | 4 | 1256 | 3605559 |