Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032536.2 | NP_115925.2 | 2 | 925 | ||
XM_006717304.3 | XP_006717367.1 | 4 | 946 | |||
XM_011519094.2 | XP_011517396.1 | 4 | 946 | |||
XM_011519096.2 | XP_011517398.1 | 4 | 1343 | |||
XM_011519097.2 | XP_011517399.1 | 3 | 864 | |||
XM_011519098.2 | XP_011517400.1 | 4 | 896 | |||
XM_011519099.2 | XP_011517401.1 | 3 | 867 | |||
XM_011519100.2 | XP_011517402.1 | 2 | 796 | |||
XM_011519102.2 | XP_011517404.1 | 3 | 811 | |||
XM_011519103.2 | XP_011517405.1 | 2 | 703 | |||
XM_011519104.2 | XP_011517406.1 | 2 | 927 | |||
XM_011519105.2 | XP_011517407.1 | 2 | 793 | |||
XM_011519106.2 | XP_011517408.1 | 4 | 946 | |||
XM_011519107.2 | XP_011517409.1 | 4 | 946 | |||
XM_011519108.2 | XP_011517410.1 | 4 | 946 | |||
XM_011519109.2 | XP_011517411.1 | 4 | 946 | |||
XM_011519110.2 | XP_011517412.1 | 4 | 946 | |||
XM_017015212.1 | XP_016870701.1 | 5 | 1050 | |||
XM_017015213.1 | XP_016870702.1 | 3 | 1253 | |||
XM_017015214.1 | XP_016870703.1 | 4 | 946 | |||
XM_017015216.1 | XP_016870705.1 | 4 | 946 | |||
GenBank mRNA | AB058760.2 | - | 2 | 925 | ||
AY358165.1 | - | 3 | 495 | |||
BC013770.1 | - | 1 | 234 | 4344617 |