Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252294.1 | NP_001239223.1 | 4 | 1162 | ||
NM_012391.2 | NP_036523.1 | 5 | 1210 | |||
XM_005248988.4 | XP_005249045.2 | 5 | 1902 | |||
GenBank mRNA | AB031549.1 | - | 5 | 1181 | ||
AF071538.1 | - | 5 | 1191 | |||
AK301543.1 | - | 4 | 1162 | |||
BC021299.2 | - | 5 | 1187 | 4546860 |