Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032561.4 | NP_115950.3 | 3 | 331 | ||
XM_005261781.1 | XP_005261838.1 | 3 | 295 | |||
XM_005261782.3 | XP_005261839.1 | 3 | 175 | |||
GenBank mRNA | AF324466.1 | - | 2 | 131 | ||
AK057349.1 | - | 3 | 381 | |||
BC031998.1 | - | 3 | 213 | 4755328 | ||
BI463610.1 | - | 3 | 336 | |||
CR456440.1 | - | 3 | 211 |