Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM185A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174092.2 | NP_001167563.1 | 2, 3 | 576 | ||
NM_001282302.1 | NP_001269231.1 | 3, 4 | 753 | |||
NM_032508.3 | NP_115897.1 | 3, 4 | 753 | |||
GenBank mRNA | AF530473.1 | - | 4, 5 | 732 | ||
AK096308.1 | - | 3, 4 | 565 | |||
AK297335.1 | - | 2, 3 | 439 | |||
BC022405.1 | - | 1, 2 | 184 | |||
BC103820.1 | - | 3, 4 | 457 | |||
BC103821.1 | - | 3, 4 | 457 | 40000939 | ||
BC103822.1 | - | 3, 4 | 457 | 40000940 | ||
BG721461.1 | - | 2, 3 | 352 | |||
BJ989805.1 | - | 1, 2 | 67 | |||
BQ224383.1 | - | 3, 4 | 544 |