Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPTG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032520.4 | NP_115909.1 | 9 | 717 | ||
XM_017023782.1 | XP_016879271.1 | 9 | 733 | |||
XM_017023783.1 | XP_016879272.1 | 7 | 605 | |||
GenBank mRNA | AF302786.1 | - | 9 | 650 | ||
AK126110.1 | - | 3 | 1873 | |||
AK312067.1 | - | 9 | 685 | |||
AY203933.1 | - | 9 | 648 | |||
BC014592.1 | - | 9 | 680 | 4824054 | ||
BC131525.1 | - | 8 | 612 | |||
BU145878.1 | - | 9 | 696 |